Product Details

SNP ID
rs150512336
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:49116288 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGATGTAGAGTTCTTGATTCCAGA[A/G]GGTCTTCCGTGATGAGGCAGTTCCG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPKOW PubMed Links

Gene Details

Gene
GPKOW
Gene Name
G-patch domain and KOW motifs
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015698.5 1028 Missense Mutation CTC,TTC L317F NP_056513.2
XM_017029415.1 1028 Missense Mutation CTC,TTC L332F XP_016884904.1

View Full Product Details