Product Details

SNP ID
rs150949392
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:30559994 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCTGTAAGTCATTATAATTCTTG[A/C]AAATACTTTTGCAAGAAGATGGAAC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CXorf21 PubMed Links

Gene Details

Gene
CXorf21
Gene Name
chromosome X open reading frame 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025159.2 757 Missense Mutation TGC,TTC C121F NP_079435.1

View Full Product Details