Product Details

SNP ID
rs151135038
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:134566625 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGTCATTGGACTTTGTCCTGAAC[C/T]GGCTGAAAACGCAGAGGTGAGGAGG
Phenotype
MIM: 300296
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00629 PubMed Links

Gene Details

Gene
LINC00629
Gene Name
long intergenic non-protein coding RNA 629
There are no transcripts associated with this gene.

Gene
PLAC1
Gene Name
placenta specific 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316887.1 344 Missense Mutation AGT,GGT S20G NP_001303816.1
NM_001316888.1 344 Missense Mutation AGT,GGT S20G NP_001303817.1
NM_001316889.1 344 Missense Mutation AGT,GGT S20G NP_001303818.1
NM_021796.3 344 Missense Mutation AGT,GGT S20G NP_068568.1
XM_011531257.2 344 Missense Mutation AGT,GGT S20G XP_011529559.1
XM_011531260.2 344 Missense Mutation AGT,GGT S20G XP_011529562.1
XM_017029236.1 344 Missense Mutation AGT,GGT S20G XP_016884725.1

View Full Product Details