Product Details
- SNP ID
-
rs1052863
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:179588875 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAAATTTAATAGTTCAGACATCTTA[A/G]ACAAGACTTGACTTTTGGGCTTCAG
- Phenotype
-
MIM: 604958
MIM: 611852
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ACTL6A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3774258] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ACTL6A
- Gene Name
- actin like 6A
- Gene
- MRPL47
- Gene Name
- mitochondrial ribosomal protein L47
View Full Product Details