Product Details
- SNP ID
-
rs1064678
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:5528582 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGATGGCATGGGGGAGGGCATACCC[T/C]TCGTAGATGGGCACAGTGTGGGTGA
- Phenotype
-
MIM: 102630
- Polymorphism
- T/C, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ACTB
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs852423] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ACTB
- Gene Name
- actin beta
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001101.3 |
585 |
Silent Mutation |
|
|
NP_001092.1 |
- Gene
- LOC221946
- Gene Name
- uncharacterized LOC221946
There are no transcripts associated with this gene.
View Full Product Details