Product Details
- SNP ID
-
rs1127430
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:7799653 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTCGGCGCTCCGGGTGTAGCTGGGG[T/G]CGCAGGTTTGGCGGGCGCGGCGGCG
- Phenotype
-
- Polymorphism
- T/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
TMEM261
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs14059] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TMEM261
- Gene Name
- transmembrane protein 261
View Full Product Details