Product Details
- SNP ID
-
rs1137004
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
16
- Location
-
Chr.1:1719362 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGAATGGCTATGATGCCTACATTT[C/T]TCTTTTCTCTTTTCATCTTTTCTGT
- Phenotype
-
MIM: 116951
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CDK11A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1137005] are located under a probe and SNP(s) [rs1059831] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CDK11A
- Gene Name
- cyclin dependent kinase 11A
- Gene
- SLC35E2
- Gene Name
- solute carrier family 35 member E2
There are no transcripts associated with this gene.
View Full Product Details