Product Details

SNP ID
rs11465394
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:7743069 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGGAGGGGGGTTTGGGGTGGCAGG[A/G]GCTGGAGAAAGAAACTGTTCTTCAT
Phenotype
MIM: 604672
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD209 PubMed Links

Gene Details

Gene
CD209
Gene Name
CD209 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144893.1 869 Silent Mutation NP_001138365.1
NM_001144894.1 869 Silent Mutation NP_001138366.1
NM_001144895.1 869 Silent Mutation NP_001138367.1
NM_001144896.1 869 Silent Mutation NP_001138368.1
NM_001144897.1 869 Silent Mutation NP_001138369.1
NM_001144899.1 869 Silent Mutation NP_001138371.1
NM_021155.3 869 Silent Mutation NP_066978.1
XM_005272472.4 869 Silent Mutation XP_005272529.1

View Full Product Details