Product Details
- SNP ID
-
rs11538653
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:30292701 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GATGGTGAAGGTTGTAGGGCAGGCC[A/G]GGGACCAGGTTCTGCATCCAAGATA
- Phenotype
-
MIM: 617034
MIM: 610020
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GATSL3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs4823086] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GATSL3
- Gene Name
- GATS protein like 3
There are no transcripts associated with this gene.
- Gene
- TBC1D10A
- Gene Name
- TBC1 domain family member 10A
View Full Product Details