Product Details

SNP ID
rs11540561
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:33266024 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCCCCTAAGGTGGACAGTAGAGC[A/G]GAATCCATTGACAAGAAGATTTCTC
Phenotype
MIM: 601497 MIM: 610900
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BAG1 PubMed Links

Gene Details

Gene
BAG1
Gene Name
BCL2 associated athanogene 1
There are no transcripts associated with this gene.

Gene
CHMP5
Gene Name
charged multivesicular body protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195536.1 284 Silent Mutation GCA,GCG A28A NP_001182465.1
NM_016410.5 284 Silent Mutation GCA,GCG A28A NP_057494.3

View Full Product Details