Product Details

SNP ID
rs11544664
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:25223379 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCTTTCATTAATGTAGTCGATAT[C/G]TGCATCATCATTATAAGGACGTCTC
Phenotype
MIM: 607090
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SYF2 PubMed Links

Gene Details

Gene
SYF2
Gene Name
SYF2 pre-mRNA splicing factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015484.4 548 Missense Mutation CAT,GAT H207D NP_056299.1
NM_207170.3 548 Missense Mutation CAT,GAT H165D NP_997053.1

View Full Product Details