Product Details
- SNP ID
-
hCV175676846
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:68042979 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCACTGTTGGCTCTCCTTGCAGTT[C/T]CTGATATGTCATCTTTAGCAGTAAG
- Phenotype
-
MIM: 600685
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
KPNA2
PubMed Links
Gene Details
- Gene
- KPNA2
- Gene Name
- karyopherin subunit alpha 2
There are no transcripts associated with this gene.
View Full Product Details