Product Details

SNP ID
rs11551392
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:37851402 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAACTGTCTCAGATTTGATTGAC[C/T]AGAAAGTGTATGAGCTACAGGCCAG
Phenotype
MIM: 613383 MIM: 613556
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKRD54 PubMed Links

Gene Details

Gene
ANKRD54
Gene Name
ankyrin repeat domain 54
There are no transcripts associated with this gene.

Gene
EIF3L
Gene Name
eukaryotic translation initiation factor 3 subunit L
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242923.1 283 Nonsense Mutation CAG,TAG Q69* NP_001229852.1
NM_016091.3 283 Nonsense Mutation CAG,TAG Q69* NP_057175.1
XM_005261625.1 283 Intron XP_005261682.1
XM_006724260.3 283 Nonsense Mutation CAG,TAG Q112* XP_006724323.1
XM_017028814.1 283 Intron XP_016884303.1
Gene
MIR658
Gene Name
microRNA 658
There are no transcripts associated with this gene.

Gene
MIR659
Gene Name
microRNA 659
There are no transcripts associated with this gene.

View Full Product Details