Product Details

SNP ID
rs11555466
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:56054442 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGATGATATAACTGGTGATGGTA[C/T]GACTTCTAATGTCCTAATCATTGGA
Phenotype
MIM: 104613 MIM: 172480 MIM: 607940
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCT6A PubMed Links

Gene Details

Gene
CCT6A
Gene Name
chaperonin containing TCP1 subunit 6A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009186.1 Intron NP_001009186.1
NM_001762.3 Intron NP_001753.1
Gene
PSPH
Gene Name
phosphoserine phosphatase
There are no transcripts associated with this gene.

Gene
SNORA15
Gene Name
small nucleolar RNA, H/ACA box 15
There are no transcripts associated with this gene.

Gene
SUMF2
Gene Name
sulfatase modifying factor 2
There are no transcripts associated with this gene.

View Full Product Details