Product Details

SNP ID
rs11569781
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:39668501 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAGCGGCGCAGACCGTAGCCCCA[A/G]TGCGGGCGCAGCCCCTGACTCGGCC
Phenotype
MIM: 611801 MIM: 171190 MIM: 607048 MIM: 604488
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PGAP3 PubMed Links

Gene Details

Gene
PGAP3
Gene Name
post-GPI attachment to proteins 3
There are no transcripts associated with this gene.

Gene
PNMT
Gene Name
phenylethanolamine N-methyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002686.4 49 Missense Mutation AAT,AGT N9S NP_002677.1
XM_011524909.2 49 Intron XP_011523211.1
Gene
STARD3
Gene Name
StAR related lipid transfer domain containing 3
There are no transcripts associated with this gene.

Gene
TCAP
Gene Name
titin-cap
There are no transcripts associated with this gene.

View Full Product Details