Product Details

SNP ID
rs11574571
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54664398 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGAGTCCTGTGACCTCAGTGCAC[A/G]GGGGGACCTACAGGTGCTTCAGCTC
Phenotype
MIM: 604821
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LILRB4 PubMed Links

Gene Details

Gene
LILRB4
Gene Name
leukocyte immunoglobulin like receptor B4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278426.3 807 Missense Mutation AGG,GGG R190G NP_001265355.2
NM_001278427.3 807 Missense Mutation AGG,GGG R190G NP_001265356.2
NM_001278428.3 807 Missense Mutation AGG,GGG R190G NP_001265357.2
NM_001278429.3 807 Missense Mutation AGG,GGG R219G NP_001265358.2
NM_001278430.3 807 Intron NP_001265359.2
XM_017026215.1 807 Missense Mutation AGG,GGG R231G XP_016881704.1
XM_017026216.1 807 Missense Mutation AGG,GGG R231G XP_016881705.1
XM_017026217.1 807 Missense Mutation AGG,GGG R231G XP_016881706.1

View Full Product Details