Product Details

SNP ID
rs270790
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54727757 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATCACTGAGGACCCCTTGCGCCTC[A/G]TTGGACAGCTCCACGATGCGGGTTC
Phenotype
MIM: 610095
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
KIR3DL3 PubMed Links

Gene Details

Gene
KIR3DL3
Gene Name
killer cell immunoglobulin like receptor, three Ig domains and long cytoplasmic tail 3
There are no transcripts associated with this gene.

View Full Product Details