Product Details

SNP ID
rs1823
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:14774803 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGCTCAGGGCGGCGTCCTGCCCAA[C/G]ATCCAGGCCGTGCTGCTGCCCAAGA
Phenotype
MIM: 615069
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C12orf60 PubMed Links

Gene Details

Gene
C12orf60
Gene Name
chromosome 12 open reading frame 60
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175874.3 468 Intron NP_787070.2
XM_005253322.4 468 Intron XP_005253379.1
XM_011520568.2 468 Intron XP_011518870.1
XM_011520569.2 468 Intron XP_011518871.1
XM_017018871.1 468 Intron XP_016874360.1
XM_017018872.1 468 Intron XP_016874361.1
XM_017018873.1 468 Intron XP_016874362.1
XM_017018874.1 468 Intron XP_016874363.1
Gene
H2AFJ
Gene Name
H2A histone family member J
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_177925.3 468 Missense Mutation AAC,AAG N111K NP_808760.1
Gene
HIST4H4
Gene Name
histone cluster 4, H4
There are no transcripts associated with this gene.

Gene
LOC105369669
Gene Name
uncharacterized LOC105369669
There are no transcripts associated with this gene.

View Full Product Details