Product Details
- SNP ID
-
rs1860871
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:150692505 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGGGAAGCTGGGGAAATAGAGAGAT[T/C]GTCATTATTGACACACCAGATATGT
- Phenotype
-
MIM: 608085
- Polymorphism
- T/C, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GIMAP2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11558054] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GIMAP2
- Gene Name
- GTPase, IMAP family member 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_015660.2 |
307 |
Silent Mutation |
ATC,ATT |
I73I |
NP_056475.1 |
View Full Product Details