Product Details

SNP ID
rs671287
Assay Type
Functionally tested
NCBI dbSNP Submissions
21
Location
Chr.1:109107946 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCGGGATCAAAATAGTAGGCATCC[C/T]GCCTCATGGAAGTAATTGTGGGGGT
Phenotype
MIM: 611298
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C1orf194 PubMed Links

Gene Details

Gene
C1orf194
Gene Name
chromosome 1 open reading frame 194
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001122961.1 663 Missense Mutation CAG,CGG Q46R NP_001116433.1
XM_005270448.3 663 Intron XP_005270505.1
XM_006710342.3 663 Missense Mutation CAG,CGG Q157R XP_006710405.1
XM_006710343.3 663 Intron XP_006710406.1
XM_011540647.2 663 Missense Mutation CAG,CGG Q167R XP_011538949.1
XM_011540648.1 663 Missense Mutation CAG,CGG Q167R XP_011538950.1
Gene
KIAA1324
Gene Name
KIAA1324
There are no transcripts associated with this gene.

Gene
SCARNA2
Gene Name
small Cajal body-specific RNA 2
There are no transcripts associated with this gene.

View Full Product Details