Product Details
- SNP ID
-
rs1044835
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:5441126 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCCTACTATCACACTGGTCTTTAAA[G/A]AAAAAAAAAAATGCATGGCCACAGT
- Phenotype
-
MIM: 601269
MIM: 614405
- Polymorphism
- G/A, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
C1QBP
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs145350626] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C1QBP
- Gene Name
- complement C1q binding protein
There are no transcripts associated with this gene.
- Gene
- DHX33
- Gene Name
- DEAH-box helicase 33
- Gene
- RPAIN
- Gene Name
- RPA interacting protein
There are no transcripts associated with this gene.
View Full Product Details