Product Details

SNP ID
rs11119
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:69452974 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGCCTCTGTCTCCGAGCTCGCCT[G/T]CATCTACTCGGCCCTCATTCTGCAC
Phenotype
MIM: 605064 MIM: 180520
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KIF23 PubMed Links

Gene Details

Gene
KIF23
Gene Name
kinesin family member 23
There are no transcripts associated with this gene.

Gene
RPLP1
Gene Name
ribosomal protein lateral stalk subunit P1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003.2 Intron NP_000994.1
NM_213725.1 Intron NP_998890.1

View Full Product Details