Product Details

SNP ID
rs11161597
Assay Type
Functionally tested
NCBI dbSNP Submissions
35
Location
Chr.1:85319253 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAGCAGAATGCAATGTAGAAGAG[A/G]CACACAGAGTCTAAGTGATTTCTCC
Phenotype
MIM: 604743
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DDAH1 PubMed Links

Gene Details

Gene
DDAH1
Gene Name
dimethylarginine dimethylaminohydrolase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134445.1 3218 UTR 3 NP_001127917.1
NM_012137.3 3218 UTR 3 NP_036269.1
XM_005270707.2 3218 UTR 3 XP_005270764.1
XM_005270710.2 3218 UTR 3 XP_005270767.1
XM_011541158.1 3218 UTR 3 XP_011539460.1
XM_017000889.1 3218 UTR 3 XP_016856378.1
XM_017000890.1 3218 UTR 3 XP_016856379.1

View Full Product Details