Product Details

SNP ID
rs11164864
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:91505921 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCTTTCACATCTGTTCTCCACTT[G/T]TCATTTTTCTGCTTAAAAACTTCCT
Phenotype
MIM: 603311
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CDC7 PubMed Links

Gene Details

Gene
CDC7
Gene Name
cell division cycle 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134419.1 Intron NP_001127891.1
NM_001134420.1 Intron NP_001127892.1
NM_003503.3 Intron NP_003494.1
XM_005271241.2 Intron XP_005271298.1
XM_005271244.2 Intron XP_005271301.1
XM_017002425.1 Intron XP_016857914.1
XM_017002426.1 Intron XP_016857915.1
XM_017002427.1 Intron XP_016857916.1

View Full Product Details