Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001242805.2 | 90 | UTR 5 | NP_001229734.2 | ||
NM_001242806.2 | 90 | UTR 5 | NP_001229735.2 | ||
NM_001242807.2 | 90 | Intron | NP_001229736.2 | ||
NM_001242808.2 | 90 | UTR 5 | NP_001229737.2 | ||
NM_001242810.2 | 90 | UTR 5 | NP_001229739.2 | ||
NM_001726.4 | 90 | Intron | NP_001717.3 | ||
NM_207189.3 | 90 | UTR 5 | NP_997072.2 | ||
XM_006710853.3 | 90 | UTR 5 | XP_006710916.1 | ||
XM_006710854.3 | 90 | UTR 5 | XP_006710917.1 | ||
XM_006710855.3 | 90 | UTR 5 | XP_006710918.1 | ||
XM_006710856.3 | 90 | Intron | XP_006710919.1 | ||
XM_006710857.3 | 90 | Intron | XP_006710920.1 | ||
XM_011542032.2 | 90 | UTR 5 | XP_011540334.1 | ||
XM_011542033.2 | 90 | Intron | XP_011540335.1 | ||
XM_011542034.2 | 90 | UTR 5 | XP_011540336.1 | ||
XM_011542035.2 | 90 | Intron | XP_011540337.1 | ||
XM_011542036.2 | 90 | Intron | XP_011540338.1 |