Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_004719.2 | 8232 | UTR 3 | NP_004710.2 | ||
XM_005269230.2 | 8232 | UTR 3 | XP_005269287.2 | ||
XM_011538984.2 | 8232 | UTR 3 | XP_011537286.1 | ||
XM_011538985.1 | 8232 | UTR 3 | XP_011537287.1 | ||
XM_017020217.1 | 8232 | UTR 3 | XP_016875706.1 | ||
XM_017020218.1 | 8232 | Intron | XP_016875707.1 | ||
XM_017020219.1 | 8232 | UTR 3 | XP_016875708.1 | ||
XM_017020220.1 | 8232 | UTR 3 | XP_016875709.1 | ||
XM_017020221.1 | 8232 | UTR 3 | XP_016875710.1 | ||
XM_017020222.1 | 8232 | UTR 3 | XP_016875711.1 | ||
XM_017020223.1 | 8232 | UTR 3 | XP_016875712.1 |