Product Details

SNP ID
rs11188570
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:96042883 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCATGGGTTTGACAAGCAAGGTTA[C/T]TAGAAGTAATCTTAAATGTCTTCAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CC2D2B PubMed Links

Gene Details

Gene
CC2D2B
Gene Name
coiled-coil and C2 domain containing 2B
There are no transcripts associated with this gene.

Gene
CCNJ
Gene Name
cyclin J
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134375.1 36 Intron NP_001127847.1
NM_001134376.1 36 Intron NP_001127848.1
NM_019084.4 36 Intron NP_061957.2
XM_005269921.3 36 Intron XP_005269978.1
XM_011539885.2 36 UTR 5 XP_011538187.1
XM_017016354.1 36 Intron XP_016871843.1
XM_017016355.1 36 Intron XP_016871844.1
XM_017016356.1 36 Intron XP_016871845.1
Gene
ENTPD1-AS1
Gene Name
ENTPD1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details