Product Details

SNP ID
rs11235740
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:73409387 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCCCAAGCCCCCTGGCACAGGAT[A/C]TCTTGTGCTGTGCTGCAGCCAATTC
Phenotype
MIM: 616316
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FAM168A PubMed Links

Gene Details

Gene
FAM168A
Gene Name
family with sequence similarity 168 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286050.1 Intron NP_001272979.1
NM_001286051.1 Intron NP_001272980.1
NM_015159.2 Intron NP_055974.1
XM_017017415.1 Intron XP_016872904.1
XM_017017416.1 Intron XP_016872905.1

View Full Product Details