Product Details

SNP ID
rs11246330
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:845077 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGTGGGATCAGAAACTGCCCGCC[C/G]GGAGGGCTGCATCCCTCCTGCCGCT
Phenotype
MIM: 602243 MIM: 601189 MIM: 602644
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CD151 PubMed Links

Gene Details

Gene
CD151
Gene Name
CD151 molecule (Raph blood group)
There are no transcripts associated with this gene.

Gene
POLR2L
Gene Name
RNA polymerase II subunit L
There are no transcripts associated with this gene.

Gene
TSPAN4
Gene Name
tetraspanin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025234.1 Intron NP_001020405.1
NM_001025235.1 Intron NP_001020406.1
NM_001025236.1 Intron NP_001020407.1
NM_001025237.1 Intron NP_001020408.1
NM_001025238.1 Intron NP_001020409.1
NM_001025239.1 Intron NP_001020410.1
NM_003271.4 Intron NP_003262.1
XM_005253102.1 Intron XP_005253159.1
XM_005253104.1 Intron XP_005253161.1
XM_011520336.1 Intron XP_011518638.1
XM_011520337.2 Intron XP_011518639.1
XM_011520338.1 Intron XP_011518640.1
XM_011520339.2 Intron XP_011518641.1
XM_011520340.2 Intron XP_011518642.1
XM_011520341.2 Intron XP_011518643.1

View Full Product Details