Product Details
- SNP ID
-
rs1131883
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:143391438 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCTTGCTCCTTGCACCCTGATTGGG[T/C]ATGGGGTGAGAGGAGGGATCAGTCC
- Phenotype
-
MIM: 179610
MIM: 602002
- Polymorphism
- T/C, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
EPHA1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1131885] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- EPHA1
- Gene Name
- EPH receptor A1
- Gene
- FAM131B
- Gene Name
- family with sequence similarity 131 member B
There are no transcripts associated with this gene.
- Gene
- MIR6892
- Gene Name
- microRNA 6892
There are no transcripts associated with this gene.
- Gene
- ZYX
- Gene Name
- zyxin
View Full Product Details