Product Details

SNP ID
rs1153104
Assay Type
Functionally tested
NCBI dbSNP Submissions
20
Location
Chr.1:1479683 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACACACCCAAACACACATGGGTCC[G/T]CAGGCACACACTCCCGCATGGGGCA
Phenotype
MIM: 612317
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ATAD3B PubMed Links

Gene Details

Gene
ATAD3B
Gene Name
ATPase family, AAA domain containing 3B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317238.1 Intron NP_001304167.1
NM_031921.5 Intron NP_114127.3
XM_005244806.3 Intron XP_005244863.1
XM_011542241.2 Intron XP_011540543.1
XM_011542243.2 Intron XP_011540545.1
XM_011542244.1 Intron XP_011540546.1
Gene
ATAD3C
Gene Name
ATPase family, AAA domain containing 3C
There are no transcripts associated with this gene.

View Full Product Details