Product Details

SNP ID
rs11541134
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:3663563 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCTCGGTGTCCAGGAGAGAAAGC[C/T]GGTCCCAGAGGCCCCAGGCCAGGGA
Phenotype
MIM: 606272 MIM: 602836 MIM: 616484
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CTNS PubMed Links

Gene Details

Gene
CTNS
Gene Name
cystinosin, lysosomal cystine transporter
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031681.2 713 Intron NP_001026851.2
NM_004937.2 713 Intron NP_004928.2
XM_005256485.2 713 Intron XP_005256542.1
XM_006721463.2 713 Intron XP_006721526.1
XM_006721464.2 713 Intron XP_006721527.1
XM_011523691.2 713 Intron XP_011521993.1
XM_011523692.2 713 Intron XP_011521994.1
XM_017024254.1 713 Intron XP_016879743.1
XM_017024255.1 713 Intron XP_016879744.1
XM_017024256.1 713 Intron XP_016879745.1
XM_017024257.1 713 Intron XP_016879746.1
XM_017024258.1 713 Intron XP_016879747.1
Gene
EMC6
Gene Name
ER membrane protein complex subunit 6
There are no transcripts associated with this gene.

Gene
P2RX5
Gene Name
purinergic receptor P2X 5
There are no transcripts associated with this gene.

Gene
P2RX5-TAX1BP3
Gene Name
P2RX5-TAX1BP3 readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
TAX1BP3
Gene Name
Tax1 binding protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204698.1 713 UTR 3 NP_001191627.1
NM_014604.3 713 UTR 3 NP_055419.1

View Full Product Details