Product Details

SNP ID
rs11542475
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:31852815 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCCAGAGACTCCACACCTACCAT[G/A]TGGATTCCCTTGATCTGCTTTCAAC
Phenotype
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
COPRS PubMed Links

Gene Details

Gene
COPRS
Gene Name
coordinator of PRMT5 and differentiation stimulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018405.3 490 Missense Mutation CAT,TAT H128Y NP_060875.2
XM_005258000.1 490 Missense Mutation CAT,TAT H116Y XP_005258057.1
Gene
UTP6
Gene Name
UTP6, small subunit processome component
There are no transcripts associated with this gene.

View Full Product Details