Product Details
- SNP ID
-
rs11539615
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:90609944 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTTTATAACCGCAGAGTAAATAAA[G/A]TATAATAGAGAATAGACTTGTAACA
- Phenotype
-
MIM: 607863
- Polymorphism
- G/A, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DIRAS2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs7854469] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DIRAS2
- Gene Name
- DIRAS family GTPase 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_017594.4 |
4274 |
UTR 3 |
|
|
NP_060064.2 |
View Full Product Details