Product Details

SNP ID
rs11545581
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:49077663 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTACCTGAGATCTCTGAGAACTTG[T/G]GACTACTACCACCGCCCACCAAGGC
Phenotype
MIM: 300947 MIM: 300840 MIM: 300526
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
CCDC120 PubMed Links

Gene Details

Gene
CCDC120
Gene Name
coiled-coil domain containing 120
There are no transcripts associated with this gene.

Gene
PRAF2
Gene Name
PRA1 domain family member 2
There are no transcripts associated with this gene.

Gene
WDR45
Gene Name
WD repeat domain 45
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001029896.1 654 Missense Mutation CAC,CCC H72P NP_001025067.1
NM_007075.3 654 Missense Mutation CAC,CCC H72P NP_009006.2

View Full Product Details