Product Details

SNP ID
rs11546682
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:23288582 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCTCAGCCGAGAATGACTTCGTCC[A/G]CCGGATCCAGGAGGAACTGGACCGC
Phenotype
MIM: 606663
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOXL2 PubMed Links

Gene Details

Gene
LOXL2
Gene Name
lysyl oxidase like 2
There are no transcripts associated with this gene.

Gene
R3HCC1
Gene Name
R3H domain and coiled-coil containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136108.2 143 Missense Mutation CAC,CGC H20R NP_001129580.2
NM_001301650.1 143 Missense Mutation CAC,CGC H20R NP_001288579.1

View Full Product Details