Product Details

SNP ID
rs11545920
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:36014859 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTGAGGCCAGTGACTAGAACGTA[G/A]ATTGAAATCGGGGTTCCAGGATTTG
Phenotype
MIM: 613304 MIM: 607382 MIM: 615535
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
ALKBH6 PubMed Links

Gene Details

Gene
ALKBH6
Gene Name
alkB homolog 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297701.1 3257 Intron NP_001284630.1
NM_032878.3 3257 Intron NP_116267.3
NM_198867.1 3257 Intron NP_942567.1
XM_005259357.4 3257 Intron XP_005259414.1
Gene
CLIP3
Gene Name
CAP-Gly domain containing linker protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199570.1 3257 UTR 3 NP_001186499.1
NM_015526.2 3257 UTR 3 NP_056341.1
Gene
LOC101927572
Gene Name
uncharacterized LOC101927572
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290056.1 3257 Intron NP_001276985.1
Gene
SYNE4
Gene Name
spectrin repeat containing nuclear envelope family member 4
There are no transcripts associated with this gene.

View Full Product Details