Product Details

SNP ID
rs11545713
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:175179145 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTACTGTTGCAGCACCTGCACCAA[A/T]AAATTTGGCAGCAGTATCAATGTCT
Phenotype
MIM: 602736
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ATP5G3 PubMed Links

Gene Details

Gene
ATP5G3
Gene Name
ATP synthase, H+ transporting, mitochondrial Fo complex subunit C3 (subunit 9)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002258.4 406 Missense Mutation ATT,TTT I76F NP_001002258.1
NM_001190329.1 406 Missense Mutation ATT,TTT I76F NP_001177258.1
NM_001689.4 406 Missense Mutation ATT,TTT I76F NP_001680.1

View Full Product Details