Product Details
- SNP ID
-
rs11546582
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:49129252 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATATGTGGTGCTTACCAGCTTGCGA[G/A]TTCGGTCCAACACAAGGTCAATGAT
- Phenotype
-
MIM: 602530
- Polymorphism
- G/A, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TUBA1B
PubMed Links
Gene Details
- Gene
- TUBA1B
- Gene Name
- tubulin alpha 1b
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006082.2 |
586 |
Missense Mutation |
ACT,ATT |
T122I |
NP_006073.2 |
View Full Product Details