Product Details

SNP ID
rs11548923
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:34753166 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATGTAGATAACTCTCCCTTAATAC[C/T]ACCACTTTAAAAAAAAATCATCAAT
Phenotype
MIM: 605680
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BAZ1A PubMed Links

Gene Details

Gene
BAZ1A
Gene Name
bromodomain adjacent to zinc finger domain 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013448.2 3708 UTR 3 NP_038476.2
NM_182648.1 3708 UTR 3 NP_872589.1
XM_011536374.1 3708 UTR 3 XP_011534676.1
XM_011536376.2 3708 UTR 3 XP_011534678.2
XM_017020942.1 3708 UTR 3 XP_016876431.1

View Full Product Details