Product Details
- SNP ID
-
rs11546960
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.21:42315334 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCAGCAGAGCTGGAGGACAGCAAG[G/A]CCAGGACCAGCCCCAGCATGCAGAG
- Phenotype
-
MIM: 600633
- Polymorphism
- G/A, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TFF3
PubMed Links
Gene Details
- Gene
- TFF3
- Gene Name
- trefoil factor 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003226.3 |
263 |
Missense Mutation |
GCC,GTC |
A28V |
NP_003217.3 |
View Full Product Details