Product Details

SNP ID
rs11548150
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:61816728 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGCGCGGGGTAGGTCCCTGAGCCG[T/C]GGTCTCGGCGGCCACCGGGTCGGGG
Phenotype
MIM: 606148 MIM: 606149
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
FADS1 PubMed Links

Gene Details

Gene
FADS1
Gene Name
fatty acid desaturase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013402.4 330 Missense Mutation ACG,GCG T68A NP_037534.3
XM_011545022.2 330 Intron XP_011543324.1
Gene
FADS2
Gene Name
fatty acid desaturase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281501.1 330 Intron NP_001268430.1
NM_001281502.1 330 Intron NP_001268431.1
NM_004265.3 330 Intron NP_004256.1
Gene
MIR1908
Gene Name
microRNA 1908
There are no transcripts associated with this gene.

View Full Product Details