Product Details
- SNP ID
-
hCV175983197
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:73537191 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTCCAGCCCGGAAGGCTCATGTGT[G/A]CAGGCTCTTGACCCCAGTGCTTGCC
- Phenotype
-
MIM: 605846
- Polymorphism
- G/A, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
BCL7B
PubMed Links
Gene Details
- Gene
- BCL7B
- Gene Name
- BCL tumor suppressor 7B
There are no transcripts associated with this gene.
View Full Product Details