Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_198834.2 | 9967 | UTR 3 | NP_942131.1 | ||
NM_198836.2 | 9967 | UTR 3 | NP_942133.1 | ||
NM_198837.1 | 9967 | UTR 3 | NP_942134.1 | ||
NM_198838.1 | 9967 | UTR 3 | NP_942135.1 | ||
NM_198839.2 | 9967 | UTR 3 | NP_942136.1 | ||
XM_005257267.4 | 9967 | UTR 3 | XP_005257324.1 | ||
XM_006721853.1 | 9967 | UTR 3 | XP_006721916.1 | ||
XM_011524701.1 | 9967 | UTR 3 | XP_011523003.1 | ||
XM_011524703.1 | 9967 | UTR 3 | XP_011523005.1 | ||
XM_011524704.2 | 9967 | UTR 3 | XP_011523006.1 | ||
XM_017024553.1 | 9967 | UTR 3 | XP_016880042.1 | ||
XM_017024554.1 | 9967 | UTR 3 | XP_016880043.1 | ||
XM_017024555.1 | 9967 | UTR 3 | XP_016880044.1 |