Product Details

SNP ID
rs11556954
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:159920360 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCCTTGAGCCAGTTCTGGAAGTT[G/C]TCGCGTCCAGGCTGGGGCCGGCCCA
Phenotype
MIM: 609738 MIM: 604634
Polymorphism
G/C, Transversion substitution
Allele Nomenclature
Literature Links
IGSF9 PubMed Links

Gene Details

Gene
IGSF9
Gene Name
immunoglobulin superfamily member 9
There are no transcripts associated with this gene.

Gene
TAGLN2
Gene Name
transgelin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277223.1 460 Intron NP_001264152.1
NM_001277224.1 460 Missense Mutation GAC,GAG D71E NP_001264153.1
NM_003564.2 460 Intron NP_003555.1

View Full Product Details