Product Details

SNP ID
rs11554800
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:128422893 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTGCCACGGACCACAATGTGGAC[A/G]ACACCACAGAGATGCTGCAGGAGTG
Phenotype
MIM: 616626
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CERCAM PubMed Links

Gene Details

Gene
CERCAM
Gene Name
cerebral endothelial cell adhesion molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286760.1 621 UTR 5 NP_001273689.1
NM_016174.4 621 Missense Mutation AAC,GAC N75D NP_057258.3
XM_005252035.1 621 Missense Mutation AAC,GAC N75D XP_005252092.1
XM_011518762.1 621 Missense Mutation AAC,GAC N75D XP_011517064.1
XM_011518763.2 621 UTR 5 XP_011517065.1
XM_017014794.1 621 Missense Mutation AAC,GAC N75D XP_016870283.1

View Full Product Details