Product Details

SNP ID
rs11550970
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:58782041 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTGTCTCTCTGTGTTATTTTTTT[C/T]CTTCGTTGCCCCTCAGCCCCCGACG
Phenotype
MIM: 165160
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
JUN PubMed Links

Gene Details

Gene
JUN
Gene Name
Jun proto-oncogene, AP-1 transcription factor subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002228.3 2073 UTR 3 NP_002219.1
Gene
LINC01135
Gene Name
long intergenic non-protein coding RNA 1135
There are no transcripts associated with this gene.

View Full Product Details