Product Details

SNP ID
rs11552121
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:21629702 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCAGCGATGAGGCGCCACGGCGG[G/A]CCGGACCCACGGGGCGGCCCTCGGA
Phenotype
MIM: 603721
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
CCDC116 PubMed Links

Gene Details

Gene
CCDC116
Gene Name
coiled-coil domain containing 116
There are no transcripts associated with this gene.

Gene
UBE2L3
Gene Name
ubiquitin conjugating enzyme E2 L3
There are no transcripts associated with this gene.

Gene
YDJC
Gene Name
YdjC homolog (bacterial)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017964.1 261 Missense Mutation GCC,GTC A75V NP_001017964.1
XM_011529906.2 261 Missense Mutation GCC,GTC A75V XP_011528208.1

View Full Product Details