Product Details

SNP ID
rs11550899
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:156738709 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCAGGGCAGAAAGACGCATGCCT[T/G]GAGGTTGTAAGAAATCCCTTTGCCA
Phenotype
MIM: 600339 MIM: 611836
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
HDGF PubMed Links

Gene Details

Gene
HDGF
Gene Name
hepatoma-derived growth factor
There are no transcripts associated with this gene.

Gene
MRPL24
Gene Name
mitochondrial ribosomal protein L24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024540.3 246 UTR 5 NP_078816.2
NM_145729.2 246 UTR 5 NP_663781.1
XM_011509981.2 246 UTR 5 XP_011508283.1
XM_011509982.2 246 UTR 5 XP_011508284.1
Gene
RRNAD1
Gene Name
ribosomal RNA adenine dimethylase domain containing 1
There are no transcripts associated with this gene.

View Full Product Details