Product Details

SNP ID
rs11558067
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:49185975 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAAAACCAAGAAGCCCTGGAGAC[T/C]CGTGCACTGGTCGGCCTATAACAAA
Phenotype
MIM: 602529
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
TUBA1A PubMed Links

Gene Details

Gene
TUBA1A
Gene Name
tubulin alpha 1a
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270399.1 824 Missense Mutation AGT,GGT S131G NP_001257328.1
NM_001270400.1 824 Missense Mutation AGT,GGT S96G NP_001257329.1
NM_006009.3 824 Missense Mutation AGT,GGT S131G NP_006000.2

View Full Product Details